Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsta4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016676.2 | NP_001016676.1 | 2 - 3 | 133 | 77 | |
XM_012963017.2 | XP_012818471.1 | 2 - 3 | 264 | 77 | ||
XM_018093890.1 | XP_017949379.1 | 3 - 4 | 294 | 77 | ||
GenBank mRNA | BC167922.1 | - | 2 - 3 | 157 | 77 | |
BC170614.1 | - | 2 - 3 | 131 | 77 | ||
BC170620.1 | - | 2 - 3 | 131 | 77 | ||
CR760229.2 | - | 2 - 3 | 133 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |