Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016811.1 | NP_001016811.1 | 3 - 4 | 920 | 67 | |
XM_012968444.2 | XP_012823898.1 | 3 - 4 | 989 | 67 | ||
GenBank mRNA | BC167578.1 | - | 3 - 4 | 940 | 67 | |
CR855567.1 | - | 3 - 4 | 920 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |