Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tcaim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016864.3 | NP_001016864.1 | 3 - 4 | 292 | 102 | |
XM_012964746.2 | XP_012820200.1 | 4 - 5 | 430 | 102 | ||
XM_012964747.2 | XP_012820201.1 | 4 - 5 | 459 | 102 | ||
XM_012964749.2 | XP_012820203.1 | 4 - 5 | 462 | 102 | ||
XM_012964750.2 | XP_012820204.1 | 4 - 5 | 440 | 102 | ||
XM_018094663.1 | XP_017950152.1 | 4 - 5 | 363 | 102 | ||
GenBank mRNA | BC123041.1 | - | 2 - 3 | 185 | 102 | |
CR848646.2 | - | 3 - 4 | 294 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |