Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016885.2 | NP_001016885.1 | 2 - 3 | 2261 | 70 | |
XM_012963041.2 | XP_012818495.1 | 3 - 4 | 2299 | 70 | ||
GenBank mRNA | BC093454.1 | - | 2 - 3 | 2261 | 70 | |
CR848579.2 | - | 2 - 3 | 2255 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |