Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qtrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016891.2 | NP_001016891.1 | 4 - 5 | 470 | 71 | |
GenBank mRNA | BC135739.1 | - | 4 - 5 | 378 | 71 | |
CR848556.2 | - | 4 - 5 | 470 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |