Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016936.2 | NP_001016936.1 | 9 - 10 | 1212 | 61 | |
GenBank mRNA | BC167123.1 | - | 8 - 9 | 1175 | 61 | |
BC167345.1 | - | 8 - 9 | 1451 | 61 | ||
CR762076.2 | - | 9 - 10 | 1212 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |