Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016949.2 | NP_001016949.1 | 5 - 6 | 1265 | 75 | |
XM_012964041.2 | XP_012819495.1 | 6 - 7 | 1385 | 75 | ||
XM_012964042.2 | XP_012819496.1 | 6 - 7 | 1488 | 75 | ||
GenBank mRNA | BC167280.1 | - | 5 - 6 | 1258 | 75 | |
CR761720.2 | - | 5 - 6 | 1265 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |