Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017011.1 | NP_001017011.1 | 5 - 6 | 341 | 83 | |
GenBank mRNA | CR762246.1 | - | 5 - 6 | 341 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |