Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017038.2 | NP_001017038.1 | 77 | 62 | ||
GenBank mRNA | BC135593.1 | - | 80 | 62 | ||
CR761940.2 | - | 77 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis |