Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017188.2 | NP_001017188.1 | 2 - 3 | 281 | 98 | |
XM_012959095.2 | XP_012814549.1 | 2 - 3 | 345 | 98 | ||
XM_018092112.1 | XP_017947601.1 | 2 - 3 | 456 | 98 | ||
GenBank mRNA | CR760622.2 | - | 2 - 3 | 281 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |