Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017205.2 | NP_001017205.1 | 5 - 6 | 849 | 114 | |
GenBank mRNA | BC135154.1 | - | 5 - 6 | 860 | 114 | |
CR760502.2 | - | 5 - 6 | 849 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |