Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017300.2 | NP_001017300.1 | 5 - 6 | 456 | 92 | |
XM_012968498.2 | XP_012823952.1 | 5 - 6 | 507 | 92 | ||
GenBank mRNA | CR855597.2 | - | 5 - 6 | 456 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |