Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsbn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017304.2 | NP_001017304.1 | 3 - 4 | 1239 | 83 | |
GenBank mRNA | CR855556.2 | - | 3 - 4 | 1239 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |