Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zdhhc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017307.2 | NP_001017307.1 | 3 - 4 | 382 | 75 | |
XM_012970476.2 | XP_012825930.1 | 3 - 4 | 542 | 75 | ||
GenBank mRNA | BC121517.1 | - | 3 - 4 | 681 | 75 | |
BC157776.1 | - | 3 - 4 | 645 | 75 | ||
BC170884.1 | - | 3 - 4 | 361 | 75 | ||
BC170886.1 | - | 3 - 4 | 361 | 75 | ||
CR855529.2 | - | 3 - 4 | 382 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |