Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapkapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017347.1 | NP_001017347.1 | 1 - 2 | 246 | 86 | |
XM_012963511.2 | XP_012818965.1 | 2 - 3 | 366 | 86 | ||
GenBank mRNA | CR761979.2 | - | 1 - 2 | 246 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |