Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030330.1 | NP_001025501.1 | 2 - 3 | 377 | 81 | |
GenBank mRNA | BC080175.1 | - | 2 - 3 | 383 | 81 | |
BC161475.1 | - | 2 - 3 | 370 | 81 | ||
CR760257.1 | - | 2 - 3 | 377 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |