Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030393.1 | NP_001025564.1 | 12 - 13 | 1406 | 112 | |
GenBank mRNA | BC090604.1 | - | 12 - 13 | 1406 | 112 | |
CT030593.1 | - | 12 - 13 | 1384 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |