Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030431.1 | NP_001025602.1 | 4 - 5 | 459 | 99 | |
XM_018092119.1 | XP_017947608.1 | 3 - 4 | 298 | 99 | ||
GenBank mRNA | BC091052.1 | - | 4 - 5 | 459 | 99 | |
CR761179.2 | - | 4 - 5 | 434 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |