Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tha1p | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030440.1 | NP_001025611.1 | 4 - 5 | 413 | 88 | |
XM_012966670.2 | XP_012822124.1 | 4 - 5 | 488 | 88 | ||
XM_012966671.1 | XP_012822125.1 | 4 - 5 | 593 | 88 | ||
GenBank mRNA | BC091069.1 | - | 4 - 5 | 413 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |