Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030456.1 | NP_001025627.1 | 4 - 5 | 782 | 72 | |
XM_012957088.2 | XP_012812542.1 | 5 - 6 | 735 | 72 | ||
GenBank mRNA | BC091576.1 | - | 4 - 5 | 782 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |