Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030469.1 | NP_001025640.1 | 4 - 5 | 1276 | 75 | |
GenBank mRNA | BC091606.1 | - | 4 - 5 | 1276 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |