Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030469.1 | NP_001025640.1 | 6 - 7 | 1346 | 70 | |
XM_012957093.2 | XP_012812547.2 | 5 - 6 | 1374 | 70 | ||
XM_018091298.1 | XP_017946787.1 | 4 - 5 | 1323 | 70 | ||
GenBank mRNA | BC091606.1 | - | 6 - 7 | 1346 | 70 | |
CR942619.2 | - | 5 - 6 | 1294 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |