Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030490.1 | NP_001025661.1 | 3 - 4 | 126 | 103 | |
GenBank mRNA | BC092368.1 | - | 3 - 4 | 126 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |