Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030503.1 | NP_001025674.1 | 8 - 9 | 1031 | 64 | |
GenBank mRNA | BC093459.1 | - | 8 - 9 | 1031 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |