Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030517.1 | NP_001025688.1 | 6 - 7 | 843 | 96 | |
XM_012959135.1 | XP_012814589.1 | 6 - 7 | 910 | 96 | ||
GenBank mRNA | BC094531.1 | - | 6 - 7 | 843 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |