Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serpina3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030522.1 | NP_001025693.1 | 5 - 6 | 1237 | 77 | |
XM_012968518.2 | XP_012823972.1 | 9 - 10 | 2551 | 77 | ||
GenBank mRNA | BC094949.1 | - | 5 - 6 | 1237 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |