Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slco1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032322.1 | NP_001027493.1 | 9 - 10 | 1804 | 108 | |
GenBank mRNA | BC096506.1 | - | 9 - 10 | 1804 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |