Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032327.1 | NP_001027498.1 | 10 - 11 | 1360 | 66 | |
XM_012954931.2 | XP_012810385.1 | 10 - 11 | 1392 | 66 | ||
GenBank mRNA | BC096519.1 | - | 10 - 11 | 1360 | 66 |
Gene Symbol: LOC101733483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018090443.1 | XP_017945932.1 | 3 - 4 | 600 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |