Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032327.1 | NP_001027498.1 | 2 - 3 | 315 | 68 | |
XM_012954931.2 | XP_012810385.1 | 2 - 3 | 347 | 68 | ||
GenBank mRNA | BC096519.1 | - | 2 - 3 | 315 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |