Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl2l11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032340.1 | NP_001027511.1 | 2 - 3 | 654 | 94 | |
XM_012963081.2 | XP_012818535.1 | 2 - 3 | 1964 | 94 | ||
GenBank mRNA | BC098324.1 | - | 2 - 3 | 654 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |