Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032341.1 | NP_001027512.1 | 4 - 5 | 1524 | 95 | |
XM_012966677.2 | XP_012822131.1 | 4 - 5 | 1679 | 95 | ||
GenBank mRNA | BC098511.1 | - | 4 - 5 | 1524 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |