Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006797.2 | NP_001006798.2 | 3 - 4 | 582 | 103 | |
GenBank mRNA | CR761007.2 | - | 3 - 4 | 582 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |