Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006797.2 | NP_001006798.2 | 4 - 5 | 692 | 68 | |
GenBank mRNA | BC076665.1 | - | 4 - 5 | 642 | 68 | |
CR761007.2 | - | 4 - 5 | 692 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |