Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007507.3 | NP_001007508.2 | 7 - 8 | 1040 | 82 | |
XM_012960904.2 | XP_012816358.1 | 7 - 8 | 1055 | 82 | ||
XM_012960906.2 | XP_012816360.1 | 5 - 6 | 693 | 82 | ||
XM_018093011.1 | XP_017948500.1 | 7 - 8 | 1074 | 82 | ||
XM_018093012.1 | XP_017948501.1 | 7 - 8 | 1074 | 82 | ||
GenBank mRNA | BC079944.1 | - | 7 - 8 | 1040 | 82 | |
CR848635.2 | - | 5 - 6 | 654 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |