Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011074.2 | NP_001011074.1 | 5 - 6 | 891 | 103 | |
XM_012956842.2 | XP_012812296.1 | 5 - 6 | 1295 | 103 | ||
GenBank mRNA | BC084459.1 | - | 5 - 6 | 834 | 103 | |
CR848150.2 | - | 5 - 6 | 891 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |