Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc27a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011348.2 | NP_001011348.2 | 7 - 8 | 1822 | 116 | |
GenBank mRNA | CR848474.2 | - | 7 - 8 | 1822 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Xenopus tropicalis Probe spans exons |