Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011385.2 | NP_001011385.2 | 3 - 4 | 935 | 88 | |
XM_018092852.1 | XP_017948341.1 | 4 - 5 | 1315 | 88 | ||
XM_018092853.1 | XP_017948342.1 | 4 - 5 | 1213 | 88 | ||
XM_018092854.1 | XP_017948343.1 | 4 - 5 | 1265 | 88 | ||
XM_018092855.1 | XP_017948344.1 | 3 - 4 | 878 | 88 | ||
GenBank mRNA | BC088568.1 | - | 3 - 4 | 858 | 88 | |
CR855420.2 | - | 3 - 4 | 935 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |