Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015902.2 | NP_001015902.1 | 1 - 2 | 449 | 66 | |
GenBank mRNA | CR926440.2 | - | 1 - 2 | 449 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis Probe spans exons |