Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpeb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015925.2 | NP_001015925.1 | 1 - 2 | 1098 | 98 | |
XM_012966552.2 | XP_012822006.1 | 1 - 2 | 1332 | 98 | ||
XM_018095384.1 | XP_017950873.1 | 2 - 3 | 1209 | 98 | ||
XM_018095385.1 | XP_017950874.1 | 2 - 3 | 977 | 98 | ||
XM_018095386.1 | XP_017950875.1 | 1 - 2 | 1333 | 98 | ||
GenBank mRNA | CR926390.2 | - | 1 - 2 | 1098 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |