Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015952.2 | NP_001015952.1 | 4 - 5 | 783 | 182 | |
GenBank mRNA | BC171182.1 | - | 3 - 4 | 632 | 182 | |
BC171184.1 | - | 3 - 4 | 632 | 182 | ||
CR848473.2 | - | 4 - 5 | 783 | 182 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |