Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015952.2 | NP_001015952.1 | 8 - 9 | 1349 | 81 | |
GenBank mRNA | BC171182.1 | - | 7 - 8 | 1198 | 81 | |
BC171184.1 | - | 7 - 8 | 1198 | 81 | ||
CR848473.2 | - | 8 - 9 | 1349 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |