Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: azin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015993.3 | NP_001015993.2 | 7 - 8 | 1031 | 81 | |
GenBank mRNA | CR848307.2 | - | 7 - 8 | 970 | 81 | |
DT426956.1 | - | 3 - 4 | 457 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Xenopus tropicalis Probe spans exons |