Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hapln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016048.2 | NP_001016048.1 | 3 - 4 | 1171 | 87 | |
XM_012958989.2 | XP_012814443.1 | 4 - 5 | 1471 | 87 | ||
GenBank mRNA | BC091610.1 | - | 4 - 5 | 860 | 87 | |
BC170561.1 | - | 3 - 4 | 1151 | 87 | ||
BC170563.1 | - | 3 - 4 | 1151 | 87 | ||
CR762351.2 | - | 3 - 4 | 1171 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |