Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016064.2 | NP_001016064.1 | 4 - 5 | 240 | 96 | |
GenBank mRNA | BC171277.1 | - | 4 - 5 | 216 | 96 | |
BC171281.1 | - | 4 - 5 | 216 | 96 | ||
CR762271.2 | - | 4 - 5 | 240 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |