Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016066.2 | NP_001016066.1 | 2 - 3 | 114 | 87 | |
GenBank mRNA | CR762262.2 | - | 2 - 3 | 114 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |