Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016070.2 | NP_001016070.1 | 1 - 2 | 182 | 68 | |
XM_012968363.2 | XP_012823817.1 | 2 - 3 | 306 | 68 | ||
XM_012968364.2 | XP_012823818.1 | 2 - 3 | 364 | 68 | ||
GenBank mRNA | BC170665.1 | - | 1 - 2 | 182 | 68 | |
BC170667.1 | - | 1 - 2 | 182 | 68 | ||
CR762251.2 | - | 1 - 2 | 182 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |