Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paqr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012962038.2 | XP_012817492.1 | 3 - 4 | 515 | 91 | |
XM_012962044.2 | XP_012817498.1 | 3 - 4 | 389 | 91 | ||
XM_012962048.2 | XP_012817502.1 | 2 - 3 | 472 | 91 | ||
GenBank mRNA | BC171300.1 | - | 2 - 3 | 276 | 91 | |
CR762035.2 | - | 2 - 3 | 277 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |