Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh3bgrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016155.2 | NP_001016155.1 | 3 - 4 | 590 | 102 | |
GenBank mRNA | BC077071.1 | - | 3 - 4 | 480 | 102 | |
BC167267.1 | - | 3 - 4 | 463 | 102 | ||
BC170711.1 | - | 3 - 4 | 523 | 102 | ||
BC170713.1 | - | 3 - 4 | 523 | 102 | ||
CR761949.2 | - | 3 - 4 | 590 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |