Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gins1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016164.2 | NP_001016164.1 | 4 - 5 | 353 | 91 | |
GenBank mRNA | BC168788.1 | - | 4 - 5 | 575 | 91 | |
BC170763.1 | - | 4 - 5 | 353 | 91 | ||
BC170767.1 | - | 4 - 5 | 353 | 91 | ||
CR761910.2 | - | 4 - 5 | 353 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |