Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016169.2 | NP_001016169.1 | 2 - 3 | 674 | 98 | |
GenBank mRNA | BC170609.1 | - | 2 - 3 | 513 | 98 | |
BC170613.1 | - | 2 - 3 | 513 | 98 | ||
CR761890.2 | - | 2 - 3 | 674 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |